Assay Details
Target Gene Details
Entrez Gene ID: | 217217 |
Gene Name: | ankyrin repeat and SOCS box-containing 16 |
Gene Aliases: |
- |
Location: |
Chr.11:102268823-102278061 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Asb16 | NM_148953.2 | 2 | 383 | NP_683755.1 |
| AF403044.1 | 2 | 383 | AAL57363.1 | |
| BC127245.1 | 2 | 379 | ||
| BC127246.1 | 2 | 382 | ||
| BC152348.1 | 2 | 383 | ||
| BC152349.1 | 2 | 383 |
More Information
Set Membership: |
|

Genomic Map