Assay Details
Target Gene Details
Entrez Gene ID: | 76884 |
Gene Name: | cytoplasmic FMR1 interacting protein 2 |
Gene Aliases: |
1500004I01Rik, 6430511D02Rik, AA930218, AU022376, Pir121, mKIAA1168 |
Location: |
Chr.11:46193849-46312859 on Build GRCm38 |
Assay Gene Location: | Within Exon 32 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyfip2 | NM_001252459.1 | 31 | 5295 | NP_001239388.1 |
| NM_001252460.1 | 31 | 5350 | NP_001239389.1 | |
| NM_133769.3 | 31 | 5058 | NP_598530.2 | |
| AB093288.2 | 30 | 4930 | BAC41472.2 | |
| AK005148.1 | 1 | 445 | BAE43189.1 | |
| AK147224.1 | 31 | 5296 | BAE27779.1 | |
| AK147586.1 | 31 | 5051 | BAE28010.1 | |
| AK147632.1 | 31 | 4992 | BAE28036.1 | |
| BC028941.1 | 3 | 1691 | AAH28941.1 |
More Information
Set Membership: |
|

Genomic Map