Assay Details
Target Gene Details
Entrez Gene ID: | 52897 |
Gene Name: | RNA binding protein, fox-1 homolog (C. elegans) 3 |
Gene Aliases: |
Fox-3, Hrnbp3, NeuN, Neuna60 |
Location: |
Chr.11:118489760-118911597 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rbfox3 | NM_001024931.2 | 15 | 2764 | NP_001020102.2 |
| NM_001039167.1 | 15 | 4302 | NP_001034256.1 | |
| NM_001039168.1 | 15 | 4158 | NP_001034257.1 | |
| NM_001285436.1 | 14 | 2865 | NP_001272365.1 | |
| NM_001285437.1 | 14 | 2705 | NP_001272366.1 | |
| NM_001285438.1 | 14 | 2570 | NP_001272367.1 | |
| AK160816.1 | 14 | 2844 | BAE36031.1 |
More Information
Set Membership: |
|

Genomic Map