Assay Details
Target Gene Details
Entrez Gene ID: | 217335 |
Gene Name: | Fas (TNFRSF6) binding factor 1 |
Gene Aliases: |
1110033G01Rik, 5930413D20, mKIAA1863 |
Location: |
Chr.11:116142285-116168270 on Build GRCm38 |
Assay Gene Location: | Within Exon 31 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fbf1 | NM_172571.3 | 31 | 4586 | NP_766159.3 |
| AF241249.1 | 29 | 3870 | AAK28326.1 | |
| AK004058.1 | 5 | 845 | BAC25065.1 | |
| BC055689.1 | 11 | 1752 | AAH55689.1 | |
| BC137794.1 | 29 | 3862 | ||
| BC144918.1 | 29 | 3859 |

Genomic Map