Assay Details
Target Gene Details
Entrez Gene ID: | 75482 |
Gene Name: | heat shock protein, alpha-crystallin-related, B9 |
Gene Aliases: |
1700007H20Rik |
Location: |
Chr.11:100713850-100714575 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hspb9 | NM_029307.1 | 1 | 614 | NP_083583.1 |
| AK005714.1 | 1 | 537 | BAB24201.1 |

Genomic Map