Assay Details
Target Gene Details
Entrez Gene ID: | 56282 |
Gene Name: | mitochondrial ribosomal protein L12 |
Gene Aliases: |
0610034O11Rik, 1500031N16Rik, MRP-L12, Rpml12 |
Location: |
Chr.11:120484669-120488754 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrpl12 | NM_027204.2 | 5 | 803 | NP_081480.2 |
| AK002757.1 | 5 | 813 | ||
| AK005328.1 | 5 | 834 | BAB23955.1 | |
| AK028168.1 | 5 | 814 | BAC25787.1 | |
| AK134816.1 | 5 | 836 | BAE22297.1 | |
| AK166969.1 | 5 | 832 | BAE39152.1 | |
| AK168337.1 | 5 | 806 | BAE40275.1 | |
| AK205740.1 | 1 | 134 | ||
| AK212489.1 | 1 | 134 | ||
| AK219774.1 | 1 | 134 | ||
| BC005712.1 | 5 | 803 | AAH05712.1 | |
| BC019565.1 | 5 | 801 | AAH19565.1 |
More Information
Set Membership: |
|

Genomic Map