Assay Details
Target Gene Details
Entrez Gene ID: | 76626 |
Gene Name: | musashi RNA-binding protein 2 |
Gene Aliases: |
1700105C15Rik, AI451722, AI563628, AW489193, Msi2h |
Location: |
Chr.11:88339382-88718267 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Msi2 | NM_054043.3 | 14 | 4731 | NP_473384.1 |
| AK037632.1 | 1 | 1847 | ||
| AK038120.1 | 1 | 1847 | ||
| AK052351.1 | 1 | 1813 | ||
| AK085831.1 | 1 | 1686 | ||
| AK086558.1 | 1 | 795 |
More Information
Set Membership: |
|

Genomic Map