Assay Details
Target Gene Details
Entrez Gene ID: | 237847 |
Gene Name: | reticulon 4 receptor-like 1 |
Gene Aliases: |
Ngr3, Ngrh2, Ngrl2 |
Location: |
Chr.11:75193993-75267769 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rtn4rl1 | NM_177708.5 | 2 | 2813 | NP_808376.1 |
| AK140456.1 | 2 | 2814 | BAE24396.1 |

Genomic Map