Assay Details
Target Gene Details
Entrez Gene ID: | 51799 |
Gene Name: | RUN domain containing 3A |
Gene Aliases: |
RPIP-8, Rap2ip, Rpip8 |
Location: |
Chr.11:102393403-102402939 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rundc3a | NM_001252347.1 | 1 | 148 | NP_001239276.1 |
| NM_016759.4 | 1 | 148 | NP_058039.1 | |
| BC046319.1 | AAH46319.1 | |||
| BY258909.1 | 1 | 120 | ||
| BY277835.1 | 1 | 148 |

Genomic Map