Assay Details
Target Gene Details
Entrez Gene ID: | 319822 |
Gene Name: | SET and MYND domain containing 4 |
Gene Aliases: |
G430029E23Rik |
Location: |
Chr.11:75348433-75405707 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smyd4 | NM_001102611.1 | 1 | 160 | NP_001096081.1 |
| AK030380.1 | BAC26933.1 | |||
| AK089959.1 | 1 | 98 | BAC41013.1 | |
| AK141484.1 | 1 | 73 | ||
| AK154507.1 | BAE32637.1 | |||
| AK166057.1 | 1 | 98 | ||
| BC095952.1 | 1 | 57 | AAH95952.1 | |
| CN693729.1 | 1 | 160 |

Genomic Map