Assay Details
Target Gene Details
Entrez Gene ID: | 319822 |
Gene Name: | SET and MYND domain containing 4 |
Gene Aliases: |
G430029E23Rik |
Location: |
Chr.11:75348433-75405707 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smyd4 | NM_001102611.1 | 5 | 1084 | NP_001096081.1 |
| AK030380.1 | 5 | 921 | BAC26933.1 | |
| AK089959.1 | 5 | 1020 | BAC41013.1 | |
| AK154507.1 | 4 | 770 | BAE32637.1 | |
| AK173306.1 | 3 | 506 | BAD32584.1 | |
| BC095952.1 | 5 | 981 | AAH95952.1 | |
| BC130220.1 | 2 | 421 |

Genomic Map