Assay Details
Target Gene Details
Entrez Gene ID: | 68385 |
Gene Name: | TLC domain containing 1 |
Gene Aliases: |
- |
Location: |
Chr.11:78176716-78180820 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tlcd1 | NM_001291235.1 | 4 | 529 | NP_001278164.1 |
| NM_001291236.1 | 4 | 1005 | NP_001278165.1 | |
| NM_001291237.1 | 5 | 713 | NP_001278166.1 | |
| NM_026708.2 | 4 | 840 | NP_080984.1 | |
| AK002265.1 | 4 | 788 | ||
| AK046577.1 | 6 | 2639 | BAE20654.1 | |
| BC005702.1 | 4 | 732 | AAH05702.1 | |
| BC039182.1 | 4 | 975 | AAH39182.1 |

Genomic Map