Assay Details
Target Gene Details
Entrez Gene ID: | 75604 |
Gene Name: | transmembrane 4 superfamily member 5 |
Gene Aliases: |
2010003F10Rik, AI323987 |
Location: |
Chr.11:70505274-70511183 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tm4sf5 | NM_029360.3 | 5 | 801 | NP_083636.2 |
| AI323987.1 | 1 | 181 | ||
| AK008064.1 | 5 | 799 | BAB25438.1 | |
| BB798656.1 | 2 | 220 | ||
| BC010782.1 | 5 | 761 | AAH10782.1 |

Genomic Map