Assay Details
Target Gene Details
Entrez Gene ID: | 217353 |
Gene Name: | transmembrane channel-like gene family 6 |
Gene Aliases: |
D11Ertd204e, EVER1 |
Location: |
Chr.11:117765985-117782193 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmc6 | NM_145439.2 | 20 | 2679 | NP_663414.3 |
| AK077671.1 | 19 | 2570 | BAC36945.1 | |
| AK090179.1 | 20 | 2634 | ||
| AK183838.1 | ||||
| AK191662.1 | ||||
| AK191892.1 | ||||
| AK191934.1 | ||||
| AK192075.1 | ||||
| AK197059.1 | 1 | 50 | ||
| AK203555.1 | ||||
| AK208336.1 | 1 | 37 | ||
| AK208842.1 | 1 | 37 | ||
| AK216268.1 | 1 | 37 | ||
| AK216409.1 | 2 | 136 | ||
| AY236497.1 | 20 | 2578 | AAP69875.1 | |
| AY263158.1 | 20 | 2604 | AAP78773.1 | |
| BC004840.1 | 20 | 2604 | AAH04840.2 | |
| BC005510.1 | 13 | 1624 | AAH05510.1 | |
| BC006956.1 | 18 | 2364 | AAH06956.1 | |
| BC013502.1 | 11 | 1406 | AAH13502.1 | |
| BC058195.1 | 20 | 2604 | AAH58195.1 | |
| BP767442.1 | 1 | 284 |

Genomic Map