Assay Details
Target Gene Details
Entrez Gene ID: | 328019 |
Gene Name: | spermatogenesis associated 32 |
Gene Aliases: |
4933400C05Rik, VAD1.2 |
Location: |
Chr.11:103208127-103218432 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spata32 | NM_177801.3 | 4 | 607 | NP_808469.1 |
| AK077068.1 | 4 | 608 | BAC36591.1 | |
| BC117072.1 | 4 | 607 | ||
| BC120775.1 | 4 | 606 | ||
| DQ118647.1 | 4 | 593 | AAZ38360.1 |

Genomic Map