Assay Details
Target Gene Details
Entrez Gene ID: | 217138 |
Gene Name: | proline rich 15-like |
Gene Aliases: |
Atad4, BC022765, E230006A12Rik |
Location: |
Chr.11:96926179-96935648 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prr15l | NM_146026.1 | 3 | 413 | NP_666138.1 |
| AK077279.1 | 2 | 386 | BAC36726.1 | |
| AK090284.1 | 2 | 414 | ||
| BC022765.1 | 3 | 413 | AAH22765.1 | |
| BC024510.1 | 2 | 297 | AAH24510.1 | |
| BC034192.1 | 2 | 343 | AAH34192.1 | |
| BC034213.1 | 2 | 392 | AAH34213.1 | |
| BC036171.1 | 2 | 360 | AAH36171.1 |

Genomic Map