Assay Details
Target Gene Details
Entrez Gene ID: | 207607 |
Gene Name: | coiled-coil domain containing 40 |
Gene Aliases: |
B930008I02Rik |
Location: |
Chr.11:119228572-119265248 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc40 | NM_175430.4 | 19 | 4190 | NP_780639.1 |
| AK046971.1 | 19 | 4193 | BAC32932.1 | |
| AK077170.1 | 4 | 852 | BAC36659.1 | |
| BC085159.1 | 21 | 3749 | AAH85159.1 |

Genomic Map