Assay Details
Target Gene Details
Entrez Gene ID: | 76380 |
Gene Name: | centrosomal protein 112 |
Gene Aliases: |
1700001M19Rik, 1700029K01Rik, 8430407H02Rik, AV043680, AV207351, Ccdc46, Macoco |
Location: |
Chr.11:108425068-108860615 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cep112 | NM_029586.2 | 1 | 125 | NP_083862.1 |
| NM_029606.3 | NP_083882.3 | |||
| NM_145688.2 | 1 | 134 | NP_663721.2 | |
| AK005611.1 | 1 | 126 | BAB24150.1 | |
| AK006504.1 | 1 | 120 | BAB24623.1 | |
| BC005723.1 | AAH05723.1 | |||
| BC048677.1 | 1 | 106 | AAH48677.1 | |
| BC086808.1 | AAH86808.1 | |||
| CA464637.1 | 1 | 136 |

Genomic Map