Assay Details
Target Gene Details
Entrez Gene ID: | 52897 |
Gene Name: | RNA binding protein, fox-1 homolog (C. elegans) 3 |
Gene Aliases: |
Fox-3, Hrnbp3, NeuN, Neuna60 |
Location: |
Chr.11:118489760-118911597 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rbfox3 | NM_001024931.2 | 5 | 771 | NP_001020102.2 |
| NM_001039167.1 | 5 | 771 | NP_001034256.1 | |
| NM_001039168.1 | 5 | 771 | NP_001034257.1 | |
| NM_001285436.1 | 4 | 731 | NP_001272365.1 | |
| NM_001285437.1 | 5 | 664 | NP_001272366.1 | |
| NM_001285438.1 | 4 | 577 | NP_001272367.1 | |
| AF229056.1 | 1 | 43 | AAL71903.1 | |
| AK032676.1 | 4 | 579 | ||
| AK049926.1 | 5 | 749 | ||
| AK079086.1 | 5 | 772 | BAC37531.1 | |
| AK086669.1 | 5 | 753 | ||
| AK160816.1 | 4 | 710 | BAE36031.1 | |
| BB623084.1 | 4 | 579 | ||
| BC054403.1 | 5 | 664 | AAH54403.1 | |
| FJ958310.1 | 1 | 185 | ||
| FJ958311.1 | 1 | 185 |
More Information
Set Membership: |
|

Genomic Map