Assay Details
Target Gene Details
Entrez Gene ID: | 258334 |
Gene Name: | olfactory receptor 1396 |
Gene Aliases: |
MOR276-2 |
Location: |
Chr.11:49111505-49119817 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Olfr1396 | NM_146337.1 | 1 | 665 | NP_666449.1 |
| BC110470.1 | 1 | 662 | ||
| BC110471.1 | 1 | 665 | AAI10472.1 |
Target Gene Details
Entrez Gene ID: | 18356 |
Gene Name: | olfactory receptor 56 |
Gene Aliases: |
IF7, MOR276-1 |
Location: |
Chr.11:49050733-49135387 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Olfr56 | NM_010999.2 | NP_035129.1 | ||
| AK053352.1 | BAC35358.1 | |||
| AK133471.1 | ||||
| BC141219.1 | ||||
| BC147437.1 |
More Information
Set Membership: |
|

Genomic Map