Assay Details
Target Gene Details
Entrez Gene ID: | 12702 |
Gene Name: | suppressor of cytokine signaling 3 |
Gene Aliases: |
Cis3, Cish3, EF-10, Ef10, SSI-3, Ssi3 |
Location: |
Chr.11:117966079-117970341 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Socs3 | NM_007707.3 | 2 | 1610 | NP_031733.1 |
| AK047165.1 | 2 | 1430 | BAC32977.1 | |
| AK139241.1 | 2 | 1431 | BAE23929.1 | |
| AK150719.1 | 2 | 1430 | BAE29797.1 | |
| AK151761.1 | 2 | 1430 | BAE30668.1 | |
| AK152441.1 | 2 | 1430 | BAE31222.1 | |
| AK152468.1 | 2 | 1430 | BAE31244.1 | |
| AK152514.1 | 2 | 1430 | BAE31277.1 | |
| AK152686.1 | 2 | 1430 | BAE31417.1 | |
| AK155097.1 | 2 | 1394 | ||
| AK157708.1 | 2 | 1416 | BAE34161.1 | |
| AK159395.1 | 2 | 1428 | BAE35049.1 | |
| AK170406.1 | 2 | 1428 | BAE41773.1 | |
| AK172399.1 | 2 | 1430 | BAE42985.1 | |
| BC052031.1 | 2 | 1406 | AAH52031.1 | |
| U72673.2 | 1 | 71 | AAB51035.1 | |
| U88328.1 | 1 | 1047 | AAB62403.1 |
More Information
Set Membership: |
|

Genomic Map