Assay Details
Target Gene Details
Entrez Gene ID: | 74341 |
Gene Name: | RIKEN cDNA G630025P09 gene |
Gene Aliases: |
4632403M07Rik |
Location: |
Chr.11:69803669-69806038 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| G630025P09Rik | NR_027913.1 | 1 | 154 | |
| AK137322.1 | 1 | 155 | ||
| AK139243.1 | 1 | 145 | ||
| AK165723.1 | 1 | 155 | BAE38353.1 |
Target Gene Details
Entrez Gene ID: | 380705 |
Gene Name: | transmembrane protein 102 |
Gene Aliases: |
4632403M07Rik, Cbap, Tmem102-ps |
Location: |
Chr.11:69803595-69805624 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem102 | NM_001033433.4 | 2 | 1714 | NP_001028605.2 |
| AK143262.1 | 2 | 1714 | BAE25328.1 | |
| CK032079.1 | 1 | 400 |

Genomic Map