Assay Details
Target Gene Details
Entrez Gene ID: | 68524 |
Gene Name: | WAS/WASL interacting protein family, member 2 |
Gene Aliases: |
1110014J05Rik, 5730509C05Rik, AA407487, Gm1176, Wich, Wire |
Location: |
Chr.11:98863598-98905578 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wipf2 | NM_197940.2 | 8 | 4882 | NP_922922.1 |
| AK044867.1 | 1 | 2025 | ||
| AK046226.1 | 1 | 3202 | ||
| AK077655.1 | 1 | 255 | ||
| BC057854.1 | 8 | 4671 | AAH57854.1 |
More Information
Set Membership: |
|

Genomic Map