Assay Details
Target Gene Details
Entrez Gene ID: | 52639 |
Gene Name: | WD repeat domain, phosphoinositide interacting 1 |
Gene Aliases: |
4930533H01Rik, AW411817, D11Ertd498e |
Location: |
Chr.11:109573521-109611389 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wipi1 | NM_145940.2 | 13 | 1523 | NP_666052.1 |
| AK159608.1 | 13 | 1552 | BAE35227.1 | |
| AK170563.1 | 13 | 1567 | BAE41884.1 | |
| BC025560.1 | 13 | 1523 | AAH25560.1 |

Genomic Map