Assay Details
Target Gene Details
Entrez Gene ID: | 58996 |
Gene Name: | Rho GTPase activating protein 23 |
Gene Aliases: |
A330041B18Rik |
Location: |
Chr.11:97404393-97502402 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgap23 | NM_021493.2 | 19 | 4305 | NP_067468.2 |
| AB041572.1 | 5 | 1340 | BAA95056.1 | |
| AK079135.1 | 3 | 1267 | BAC37556.1 | |
| AK173187.1 | 10 | 1951 | BAD32465.1 |

Genomic Map