Assay Details
Target Gene Details
Entrez Gene ID: | 58996 |
Gene Name: | Rho GTPase activating protein 23 |
Gene Aliases: |
A330041B18Rik |
Location: |
Chr.11:97404393-97502402 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgap23 | NM_021493.2 | 2 | 776 | NP_067468.2 |
| BU505571.1 | 2 | 662 | ||
| BU708895.1 |

Genomic Map