Assay Details
Target Gene Details
Entrez Gene ID: | 71276 |
Gene Name: | coiled-coil domain containing 57 |
Gene Aliases: |
4933434G05Rik |
Location: |
Chr.11:120826529-120932902 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc57 | NM_027745.1 | 19 | 3270 | NP_082021.1 |
| AK017054.1 | 4 | 737 | BAB30570.1 | |
| BC056480.1 | 19 | 3270 | AAH56480.1 |

Genomic Map