Assay Details
Target Gene Details
Entrez Gene ID: | 216613 |
Gene Name: | coiled-coil domain containing 85A |
Gene Aliases: |
- |
Location: |
Chr.11:28385684-28584441 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc85a | NM_001166661.2 | 1 | 872 | NP_001160133.1 |
| NM_181577.5 | 1 | 872 | NP_853555.2 | |
| AK045568.1 | 1 | 845 | BAC32419.1 | |
| AK087049.1 | 1 | 815 | BAC39790.1 | |
| AK087070.1 | 1 | 815 | BAC39795.1 | |
| AK173298.1 | 1 | 878 | BAD32576.1 | |
| BC066065.1 | 1 | 395 | AAH66065.1 |

Genomic Map