Assay Details
Target Gene Details
Entrez Gene ID: | 14252 |
Gene Name: | flotillin 2 |
Gene Aliases: |
AI573412, Esa, reggie-2 |
Location: |
Chr.11:78037868-78060434 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Flot2 | NM_001040403.1 | 11 | 2197 | NP_001035493.1 |
| NM_001284227.1 | 12 | 2281 | NP_001271156.1 | |
| NM_001284228.1 | 12 | 2144 | NP_001271157.1 | |
| NM_008028.2 | 12 | 2288 | NP_032054.1 | |
| AK004589.1 | 11 | 2135 | BAB23392.1 | |
| AK030199.1 | 11 | 2160 | BAC26837.1 | |
| AK149530.1 | 12 | 2219 | BAE28942.1 | |
| AK170557.1 | 11 | 2198 | BAE41879.1 | |
| AK179634.1 | 1 | 76 | ||
| AK180756.1 | 1 | 88 | ||
| AK181207.1 | 1 | 82 | ||
| AK182347.1 | 1 | 79 | ||
| AK183299.1 | 1 | 74 | ||
| AK193346.1 | ||||
| AK198617.1 | 1 | 281 | ||
| AK205894.1 | 1 | 46 | ||
| AK211590.1 | 1 | 281 | ||
| AK212851.1 | 1 | 281 | ||
| BC070423.1 | 12 | 2246 | AAH70423.1 | |
| U07890.1 | 12 | 2158 | AAA93127.1 |

Genomic Map