Assay Details
Target Gene Details
Entrez Gene ID: | 19062 |
Gene Name: | inositol polyphosphate 5-phosphatase K |
Gene Aliases: |
C62, Pps, SKIP |
Location: |
Chr.11:75630507-75648865 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Inpp5k | NM_008916.2 | 12 | 2431 | NP_032942.1 |
| AK052817.1 | 11 | 3175 | ||
| AK054436.1 | 12 | 2450 | BAC35778.1 | |
| AK164290.1 | 12 | 2420 | BAE37720.1 | |
| BC014725.1 | 12 | 2361 | ||
| BC066112.1 | 12 | 2431 | AAH66112.1 | |
| U96724.1 | 12 | 2462 | AAC53265.1 |
More Information
Set Membership: |
|

Genomic Map