Assay Details
Target Gene Details
Entrez Gene ID: | 26398 |
Gene Name: | mitogen-activated protein kinase kinase 4 |
Gene Aliases: |
JNKK1, MEK4, MKK4, PRKMK4, Sek1, Serk1 |
Location: |
Chr.11:65688244-65788354 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Map2k4 | NM_009157.4 | 11 | 2422 | NP_033183.1 |
| AK036928.1 | 11 | 2426 | BAC29642.1 | |
| AK044534.1 | 11 | 2414 | BAC31968.1 | |
| AK044698.1 | 11 | 2414 | BAC32038.1 | |
| AK139777.1 | 12 | 2445 | ||
| BC029833.1 | 9 | 2173 | AAH29833.1 |
More Information
Set Membership: |
|

Genomic Map