Assay Details
Target Gene Details
Entrez Gene ID: | 19084 |
Gene Name: | protein kinase, cAMP dependent regulatory, type I, alpha |
Gene Aliases: |
1300018C22Rik, RIalpha, Tse-1, Tse1 |
Location: |
Chr.11:109649395-109669659 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prkar1a | NM_021880.2 | 11 | 2214 | NP_068680.1 |
| AK005039.1 | 11 | 2216 | BAB23766.1 | |
| AK019147.1 | 9 | 1849 | BAB31568.2 | |
| AK027916.1 | 11 | 2181 | BAC25664.1 | |
| AK049832.1 | 11 | 2208 | ||
| AK051068.1 | 11 | 2215 | BAC34516.1 | |
| AK053918.1 | 11 | 2216 | ||
| AK083383.1 | 11 | 2195 | BAC38895.1 | |
| AK145797.1 | 11 | 2215 | BAE26655.1 | |
| AK145860.1 | 11 | 2182 | BAE26704.1 | |
| AK150927.1 | 10 | 1655 | BAE29965.1 | |
| AK151040.1 | 10 | 1655 | BAE30057.1 | |
| AK151556.1 | 10 | 1655 | BAE30501.1 | |
| AK152044.1 | 10 | 1655 | BAE30901.1 | |
| BC003461.1 | 11 | 2185 | AAH03461.1 |
More Information
Set Membership: |
|

Genomic Map