Assay Details
Target Gene Details
Entrez Gene ID: | 387140 |
Gene Name: | microRNA 21a |
Gene Aliases: |
Mir21, Mirn21, mmu-mir-21, mmu-mir-21a |
Location: |
Chr.11:86584067-86584158 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mir21a | NR_029738.1 | 1 | 40 | |
| AJ459711.1 |
Target Gene Details
Entrez Gene ID: | 75909 |
Gene Name: | vacuole membrane protein 1 |
Gene Aliases: |
3110098I04Rik, 4930579A11Rik, AI787464, Tango5, Tmem49, mir-21a, ni-2 |
Location: |
Chr.11:86583852-86683822 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vmp1 | NM_029478.3 | 12 | 2519 | NP_083754.2 |
| AK043091.1 | 12 | 2520 | BAC31456.1 |
More Information
Set Membership: |
|

Genomic Map