Assay Details
Target Gene Details
Entrez Gene ID: | 78919 |
Gene Name: | fibronectin type III domain containing 8 |
Gene Aliases: |
4930466G16Rik |
Location: |
Chr.11:82892145-82900737 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fndc8 | NM_030224.1 | 1 | 206 | NP_084500.1 |
| AK019639.1 | 1 | 228 | BAB31821.1 | |
| BC120835.1 | 1 | 206 | AAI20836.1 | |
| BC137682.1 | 1 | 206 |

Genomic Map