Assay Details
Target Gene Details
Entrez Gene ID: | 14233 |
Gene Name: | forkhead box I1 |
Gene Aliases: |
5830401E05Rik, FREAC6, Fkh10, HFH-3, Hfh3 |
Location: |
Chr.11:34204341-34208089 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Foxi1 | NM_023907.4 | 2 | 1205 | NP_076396.3 |
| AK019994.1 | 2 | 1207 | BAB31957.2 | |
| BC007475.1 | 2 | 1170 |
More Information
Set Membership: |
|

Genomic Map