Assay Details
Target Gene Details
Entrez Gene ID: | 19192 |
Gene Name: | proteaseome (prosome, macropain) activator subunit 3 (PA28 gamma, Ki) |
Gene Aliases: |
AA410043, AU020960, Ki, PA28gamma, REGgamma, pa28g |
Location: |
Chr.11:101316251-101323531 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Psme3 | NM_011192.3 | 11 | 1752 | NP_035322.1 |
| AK140597.1 | 11 | 1773 | BAE24425.1 | |
| AK182317.1 | 1 | 256 | ||
| AK216469.1 | 1 | 256 | ||
| AK219671.1 | 1 | 158 | ||
| BC005524.1 | 3 | 1005 | AAH05524.1 | |
| BC015255.1 | 11 | 1752 | AAH15255.1 | |
| BC139818.1 | 11 | 1700 | ||
| D87911.1 | 11 | 1675 | BAA22041.1 |

Genomic Map