Assay Details
Target Gene Details
Entrez Gene ID: | 77371 |
Gene Name: | Sec24 related gene family, member A (S. cerevisiae) |
Gene Aliases: |
- |
Location: |
Chr.11:51692263-51763627 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sec24a | NM_001290785.1 | 23 | 4319 | NP_001277714.1 |
| NM_175255.3 | 23 | 4322 | NP_780464.2 | |
| AK090341.1 | 2 | 563 | BAC41177.1 |
More Information
Set Membership: |
|

Genomic Map