Assay Details
Target Gene Details
Entrez Gene ID: | 11431 |
Gene Name: | acid phosphatase 1, soluble |
Gene Aliases: |
4632432E04Rik, AI427468, Acp-1, LMW-PTP |
Location: |
Chr.12:30893326-30911621 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acp1 | NM_001110239.1 | 6 | 1796 | NP_001103709.1 |
| NM_021330.4 | 6 | 1796 | NP_067305.2 | |
| AK014603.1 | 6 | 1767 | BAB29458.1 | |
| AK075647.1 | 1 | 238 | ||
| AK082955.1 | 6 | 1741 | BAC38707.1 | |
| BC111893.1 | 6 | 1735 | ||
| BC112413.1 | 6 | 1735 |
More Information
Set Membership: |
|

Genomic Map