Assay Details
Target Gene Details
Entrez Gene ID: | 66204 |
Gene Name: | acylphosphatase 1, erythrocyte (common) type |
Gene Aliases: |
1110039O14Rik, AI325944 |
Location: |
Chr.12:85272398-85288196 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acyp1 | NM_025421.2 | 3 | 368 | NP_079697.1 |
| AK004176.1 | 3 | 368 | BAB23208.1 | |
| AK007244.1 | 3 | 350 | BAB24911.1 | |
| AK018832.1 | 3 | 318 | BAB31454.1 | |
| BC061088.1 | 3 | 339 | ||
| BC096417.1 | 3 | 341 | AAH96417.1 | |
| BC116781.1 | 3 | 321 | ||
| BC116783.1 | 3 | 321 |
More Information
Set Membership: |
|

Genomic Map