Assay Details
Target Gene Details
Entrez Gene ID: | 100038666 |
Gene Name: | predicted gene 12298 |
Gene Aliases: |
OTTMUSG00000005912 |
Location: |
Chr.11:66905632-66947086 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm12298 | NR_033539.1 | |||
| AK131836.1 | BAE20824.1 |
Target Gene Details
Entrez Gene ID: | 193003 |
Gene Name: | phosphoinositide-interacting regulator of transient receptor potential channels |
Gene Aliases: |
A530088H08Rik |
Location: |
Chr.11:66911910-66929877 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pirt | NM_178656.3 | 2 | 2064 | NP_848771.1 |
| AK041182.1 | 2 | 2065 | BAC30853.1 | |
| AK051158.1 | 2 | 2011 | ||
| AK051285.1 | 2 | 2074 | BAC34593.1 | |
| AK148754.1 | 1 | 68 | BAE28657.1 | |
| EU447173.1 | 2 | 2055 |

Genomic Map