Assay Details
Target Gene Details
Entrez Gene ID: | 20877 |
Gene Name: | aurora kinase B |
Gene Aliases: |
AIM-1, AIRK2, AL022959, Aik2, Aim1, Ark2, AurB, IPL1, STK-1, Stk12, Stk5 |
Location: |
Chr.11:69045625-69051664 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aurkb | NM_011496.1 | 1 | 304 | NP_035626.1 |
| AK075951.1 | 1 | 270 | BAC36078.1 | |
| AK132006.1 | 1 | 298 | BAE20935.1 | |
| AK191945.1 | 1 | 163 | ||
| BC003261.1 | 1 | 296 | AAH03261.1 | |
| BY005259.1 | 1 | 311 | ||
| D21099.1 | 1 | 169 | BAA04658.1 |
More Information
Set Membership: |
|

Genomic Map