Assay Details
Target Gene Details
Entrez Gene ID: | 216560 |
Gene Name: | WD repeat containing planar cell polarity effector |
Gene Aliases: |
AV249152 |
Location: |
Chr.11:21572281-21898687 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wdpcp | NM_145425.3 | 10 | 1250 | NP_663400.2 |
| AF424701.1 | 10 | 1213 | AAL24810.1 | |
| AK043672.1 | 9 | 1137 | BAC31614.1 | |
| AK083042.1 | 10 | 1251 | BAC38742.1 | |
| BC056139.1 | AAH56139.1 | |||
| BC141166.1 | 10 | 1214 |

Genomic Map