Assay Details
Target Gene Details
Entrez Gene ID: | 238021 |
Gene Name: | fascin homolog 2, actin-bundling protein, retinal (Strongylocentrotus purpuratus) |
Gene Aliases: |
A930022G03, Ahl8, C630046B20Rik |
Location: |
Chr.11:120359851-120368173 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fscn2 | NM_172802.4 | 1 | 837 | NP_766390.2 |
| AK044556.1 | ||||
| AK050002.1 | 1 | 838 | BAC34026.1 | |
| AK078674.1 | 1 | 865 | ||
| BC109356.1 | 1 | 798 | AAI09357.1 | |
| BC109357.1 | 1 | 798 | AAI09358.1 |
More Information
Set Membership: |
|

Genomic Map