Assay Details
Target Gene Details
Entrez Gene ID: | 23879 |
Gene Name: | fragile X mental retardation, autosomal homolog 2 |
Gene Aliases: |
Fxr2h |
Location: |
Chr.11:69632971-69653297 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fxr2 | NM_011814.2 | 1 | 355 | NP_035944.2 |
| AK154243.1 | 1 | 354 | BAE32459.1 | |
| AK172042.1 | 1 | 368 | BAE42794.1 | |
| BC057007.1 | 1 | 250 | AAH57007.1 | |
| BC062971.1 | 1 | 355 | AAH62971.1 |
More Information
Set Membership: |
|

Genomic Map