Assay Details
Target Gene Details
Entrez Gene ID: | 19285 |
Gene Name: | polymerase I and transcript release factor |
Gene Aliases: |
2310075E07Rik, AW546441, Cav-p60, Cavin |
Location: |
Chr.11:100956736-100971027 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptrf | NM_008986.2 | 1 | 320 | NP_033012.1 |
| AF036249.1 | 1 | 275 | AAC53588.1 | |
| AK009381.1 | 1 | 411 | ||
| AK154132.1 | 1 | 257 | BAE32397.1 | |
| AK169463.1 | 1 | 319 | BAE41188.1 | |
| BC110698.1 | 1 | 302 | AAI10699.1 | |
| BC116701.1 | 1 | 262 | ||
| BC117527.1 | 1 | 262 | ||
| BY132697.1 | 1 | 320 |
More Information
Set Membership: |
|

Genomic Map