Assay Details
Target Gene Details
Entrez Gene ID: | 20362 |
Gene Name: | septin 8 |
Gene Aliases: |
AW046166, Sepl, mKIAA0202 |
Location: |
Chr.11:53519736-53549600 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sept8 | NM_001252332.1 | 10 | 3425 | NP_001239261.1 |
| NM_001252333.1 | 10 | 3419 | NP_001239262.1 | |
| NM_033144.2 | 10 | 3605 | NP_149156.1 | |
| AB093215.1 | 10 | 3275 | BAC41399.1 | |
| AF179996.1 | 9 | 3121 | AAG09408.1 | |
| BC049819.1 | 10 | 3240 | AAH49819.1 | |
| BC059248.1 | 10 | 3541 | AAH59248.1 |
More Information
Set Membership: |
|

Genomic Map