Assay Details
Target Gene Details
Entrez Gene ID: | 77864 |
Gene Name: | yippee-like 2 (Drosophila) |
Gene Aliases: |
6430570G24, E130113K08Rik |
Location: |
Chr.11:86936425-86993849 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ypel2 | NM_001005341.3 | 5 | 1359 | NP_001005341.1 |
| BC121190.1 | 5 | 1231 |

Genomic Map