Assay Details
Target Gene Details
Entrez Gene ID: | 268515 |
Gene Name: | BAH domain and coiled-coil containing 1 |
Gene Aliases: |
B930044J06, BC060615 |
Location: |
Chr.11:120232904-120292297 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bahcc1 | NM_198423.3 | 16 | 5255 | NP_940815.3 |
| AB257855.1 | 15 | 5257 | ||
| AK147250.1 | 16 | 5255 | BAE27797.1 | |
| AK147400.1 | 8 | 2163 | BAE27888.1 | |
| BC072602.1 | 5 | 693 | AAH72602.1 |
More Information
Set Membership: |
|

Genomic Map