Assay Details
Target Gene Details
Entrez Gene ID: | 19014 |
Gene Name: | mediator complex subunit 1 |
Gene Aliases: |
AI480703, CRSP210, DRIP205, PBP, Pparbp, TRAP220, l11Jus15 |
Location: |
Chr.11:98152154-98193314 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Med1 | NM_134027.2 | 18 | 2019 | NP_598788.2 |
| AK135480.1 | 1 | 654 | ||
| AK147199.1 | 18 | 1996 | BAE27757.1 | |
| BC021440.1 | AAH21440.1 | |||
| BC079636.1 | 18 | 1989 | AAH79636.1 |
More Information
Set Membership: |
|

Genomic Map