Assay Details
Target Gene Details
Entrez Gene ID: | 17285 |
Gene Name: | mesenchyme homeobox 1 |
Gene Aliases: |
AI385561, D330041M02Rik, Mox-1, Mox1 |
Location: |
Chr.11:101877510-101894354 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Meox1 | NM_010791.3 | 3 | 1597 | NP_034921.1 |
| AK052370.1 | 3 | 1607 | BAC34961.1 | |
| AK133672.1 | 3 | 968 | BAE21777.1 | |
| BC011082.1 | 3 | 1597 | AAH11082.1 | |
| Z15103.1 | 3 | 1552 | CAA78812.1 |
More Information
Set Membership: |
|

Genomic Map